N-Undecylphosphocholine ≥99.5%, Crystallization grade
Supplier: CUBE BIOTECH
16045
16046
16047
77392-374EA
389.98
CAD
77392-374
77392-376
77392-378
N-Undecylphosphocholine ≥99.5%, Crystallization grade
N-Undecylphosphocholine
Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.
Molecular Weight: 337.4 g/mol
Form: White crystalline powder
CMC (in H2O): 1.85 mM (0.062%)
Synonyms are: Undecylphosphocholine, Fos-11
CAS Number:
253678-65-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...