Decyl (2-(trimethylammonio)ethyl) phosphate ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

16041 16042 16043
77392-368PK 341.69 CAD
77392-368 77392-370 77392-372
Decyl (2-(trimethylammonio)ethyl) phosphate ≥99.5%, Crystallization grade
Decyl (2-(trimethylammonio)ethyl) phosphate

Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.


Molecular Weight: 323.41 g/mol
Form: White crystalline powder
CMC (in H2O): 11 mM (0.35%)
Synonyms are: Decylphosphocholine, Fos-10, n-Decylphosphocholine

Formula: C₁₅H₃₄NO₄P
MDL Number: MFCD07784458
CAS Number: 70504-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5980f04e2cb5585f77e69ad3da5f85b51eccf48&vid=3e0648f82c9fad8e56a4446f3461279df66bd1ca&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR