Miltefosine ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

16037 16038 16039
77392-362EA 372.56 CAD
77392-362 77392-364 77392-366
Miltefosine ≥99.5%, Crystallization grade
Miltefosine

Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.


Molecular Weight: 407.57 g/mol
Form: White crystalline powder
CMC (in H2O): 0.013 mM (0.00053%)
Synonyms are: Hexadecylphosphocholine, Fos-16, n-Hexadecylphosphocholine, Miltefosine

Formula: C₂₁H₄₆NO₄P
MDL Number: MFCD00133396
CAS Number: 58066-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b330ff885eef253315920456d4f73eabb02830e&vid=d23b3d97507ea7617fe877980f1132fb70f9a94f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR