N-Tridecylphosphocholine ≥99.5%, Crystallization grade
Supplier: CUBE BIOTECH
16025
16026
16027
77392-350PK
389.98
CAD
77392-350
77392-352
77392-354
N-Tridecylphosphocholine ≥99.5%, Crystallization grade
N-Tridecylphosphocholine
Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.
Molecular Weight: 365.5 g/mol
Form: White crystalline powder
CMC (in H2O): 0.75 mM (0.027%)
Synonyms are: Tridecylphosphocholine, Fos-13
CAS Number:
85775-42-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...