N-Dodecylphosphocholine ≥99.5%, Crystallization grade
Supplier: CUBE BIOTECH
16021
16022
16023
16024
77392-342EA
339.76
CAD
77392-342
77392-344
77392-346
77392-348
N-Dodecylphosphocholine ≥99.5%, Crystallization grade
N-Dodecylphosphocholine
Phosphocholines, or Fos-cholines, are a novel class of detergents that is well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer Fos-cholines with chain lengths from C-9 to C-16.
Molecular Weight: 351.46 g/mol
Form: White crystalline powder
CMC (in H2O): 1.5 mM (0.047%)
Synonyms are: Dodecylphosphocholine, Fos-12 or n-Dodecylphosphocholine
CAS Number:
29557-91-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...