n-Dodecyl-β-D-maltoside ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside

16015 16016 16014
77392-332PK 3220.01 CAD
77392-332 77392-334 77392-330
n-Dodecyl-β-D-maltoside ≥99.5%, Crystallization grade
n-Dodecyl-β-D-maltoside

Mild, non-ionic detergents such as Dodecylmaltoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer maltosides with chain lengths from C-9 to C-13.


Molecular Weight: 510.63 g/mol
Form: White crystalline powder
CMC (in H2O): 0.17 mM (0.0087%)
Mild, non-ionic detergent

Formula: C₂₄H₄₆O₁₁
MDL Number: MFCD00043012
CAS Number: 69227-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR