n-Decyl-β-D-maltoside ≥99.5%, Crystallization grade
Supplier: CUBE BIOTECH
16010
16011
16012
77392-324PK
696.91
CAD
77392-324
77392-326
77392-328
n-Decyl-β-D-maltoside ≥99.5%, Crystallization grade
n-Decyl-β-D-maltoside
Decylmaltoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer maltosides with chain lengths from C-9 to C-13.
Molecular Weight: 487.57 g/mol
Form: White crystalline powder
CMC (in H2O): 1.66 mM (0.08%)
Mild, non-ionic detergent
Formula:
C₂₂H₄₂O₁₁ |
MDL Number:
MFCD00061624 CAS Number: 82494-09-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...