N,N-Dimethyl-1-dodecylamine N-oxide ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

16006 16007 16008
77392-318PK 449.79 CAD
77392-318 77392-320 77392-322
N,N-Dimethyl-1-dodecylamine N-oxide ≥99.5%, Crystallization grade
N,N-Dimethyl-1-dodecylamine N-oxide

LDAO is one of the classic detergents that we offer in crystallization-grade quality at an attractive price. We aliquot our detergents so that they are convenient to use, keep fresh and provide optimal performance.


Molecular Weight: 229.40 g/mol
Form: White crystalline powder
CMC (in H2O): 1-2 mM (0.023%)
Classic detergent in crystallization-grade quality

Formula: C₁₄H₃₁NO
MDL Number: MFCD00002049
CAS Number: 1643-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fda5f47360000d1693286dcbde1b8cce848baac3&vid=571e9c0d6d3ee7396bcf3b69755ccdf116ec2cec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR