Mouse Recombinant MSP1D1-His (from E. coli)
Supplier: CUBE BIOTECH
26516 26512
77392-772PK 1872.55 CAD
77392-772 77392-770
Mouse Recombinant MSP1D1-His (from E. coli)
Proteins and Peptides
Membrane scaffold protein, mouse origin.
  • Extinction coefficient (in water) ε280: 25,440 M⁻¹cm⁻¹ (Human origin), 22,460 M⁻¹cm⁻¹ (Mouse origin), 33,460 M⁻¹cm⁻¹ (Rat origin), 41,940 M⁻¹cm⁻¹ (Alpaca origin)
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Affinity tag: N-Terminal his-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 (his-tagged) creates nanodiscs of 9 to 10 nm in diameter. This product contains a his-tag, keep that in mind when choosing the affinity tag for your membrane protein.
This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.