DIBMA glucosamine HEPES, PureCube Protein Solubilisation

Supplier: CUBE BIOTECH

18041 18061 18062
77392-484PK 527.28 CAD
77392-484 77392-492 77392-494
DIBMA glucosamine HEPES, PureCube Protein Solubilisation
DIBMA Glucosamine

With diisobutylene-maleic acid (DIBMA), you can directly extract membrane proteins from cells without an intermediate step of detergent solubilization, like with SDS, which would usually interfere with the protein's function. Another advantage of DIBMA as a tool for protein solubilization is the lack of an absorbance maxima at 280 nm. In comparison to SMAs, this would usually interfere with protein quantification, as aromatic amino acids absorb at the same spectrum. PureCube DIBMA is lyophilized from two different buffer solutions (HEPES or TRIS) to ensure a stable pH of 7.5, which is ideal for most protein solubilizations. A good publication to read even more details about DIBMA is Oluwole et al. 2017.


Usage: Protein solubilization
Formula Weight 10,000 g/mol
pH: 7.5 in buffer
dn/dc: 1.35 M-1
Solubility: > 10 % in H2O
Absorbance at 280 nm: < 0.3 (1 % solution)
Mg2+ Tolerance: >50 mM
Ca2+ Tolerance: >50 mM


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR