PureCube Cu-NTA MagBeads
Supplier: CUBE BIOTECH
PureCube Cu-NTA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.
- Usage: Specific binding and purification of 6× His-tagged proteins, binding of copper-binding proteins
- Specificity: Affinity to His-tagged proteins, Affinity to Copper-finger proteins
- Binding capacity: >80 mg/ml
- Bead ligand: Cu-NTA
- Bead size: 30 μm
- Filling quantity: Delivered as a 25% suspension
- Metal ion capacity: >15 µeqv Zn²⁺/ml
- pH stability: 2 to 14
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
Our year-long experience in manufacturing agarose resin lead to the high yield of 50 mg protein per ml resin, which is leading in the market compared to other Cu-NTA suppliers.
Cu-NTA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin. For your convenience Cube Biotech also offers its own MagBead separator for use with our magnetic beads.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...