PureCube Al-NTA MagBeads

Supplier: CUBE BIOTECH

31501-AL 31505-AL 31525-AL 31590-AL 31591-AL
77393-558EA 202.65 CAD
77393-558 77393-570 77393-582 77393-594 77393-606
PureCube Al-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube Al-NTA MagBeads/Magnetic beads were developed for the enrichment of phosphorylated proteins. They can also be used for the purification of His-tagged proteins as a secondary function.


  • Usage: Enrichment of phosphopeptides
  • Specificity: Affinity to phosphorylated biomolecules
  • Bead ligand: Al-NTA
  • Bead size: 30 µm
  • Filling quantity: Delivered as a 25% suspension
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The affinity matrix is based on 6% cross-linked agarose that was magnetized. The bead ligand is Al-NTA and the particle diameter is 30 µm. They are very homogeneous in size, yielding in a high degree of reproducibility. The phosphopeptide enrichment with Cube Biotech MagBeads can be automated.


As an alternative we also offer PureCube Fe-NTA Agarose resin and said resin pre-packed in cartridges/columns.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR