PureCube NHS Activated MagBeads XL
Supplier: CUBE BIOTECH
PureCube NHS (N-hydroxy succinimide) MagBeads/magnetic beads XL are your best option to couple biomolecules carrying free-standing amine groups.
- Usage: Coupling proteins to active NHS Groups
- Specificity: Exposed amine (NH₂) groups on biomolecule
- Binding capacity: >25 mg/ml
- Bead ligand: Chemically active NHS groups
- Bead size: 90 μm
- Filling quantity: Delivered as a 25% suspension
- Storage buffer: 100% isopropanol
- Matrix: 6% cross-linked agarose
- Storage temperature: 4 °C
- Shelf life: If properly stored - six months upon delivery, unopened
- Chemical structure: NHS structure
- Special notes: This product is shipped as a flammable good, 2-propanol UN1219
The resulting Amide bond between the NHS MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube NHS Activated MagBeads/magnetic beads are also available as agarose resin for FPLC or as MagBeads with a smaller bead size for more mechanical stability.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...