PureCube NTA XL Agarose

Supplier: CUBE BIOTECH

55803 55805 55810 55812
77393-290EA 362.93 CAD
77393-290 77393-292 77393-294 77393-296
PureCube NTA XL Agarose
Protein Purification Protein Purification Reagents

PureCube NTA XL (nitrilotriacetic acid) agarose resin was developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead ligand: NTA
  • Bead size: 400 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 400 µm. This size is relatively large, which is helpful for very high flow rates in, e.g., FPLC assays or very viscous cell media. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA agarose resin uncoupled to any metal ion for the case that we do not offer your desired NTA + metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR