PureCube Maleimide Activated MagBeads

Supplier: CUBE BIOTECH

51201 51205 51225 51290
77393-232EA 673.52 CAD
77393-232 77393-234 77393-236 77393-238
PureCube Maleimide Activated MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube Maleimide MagBeads are your best option to couple biomolecules that carry thiol (SH) groups.


  • Usage: Coupling proteins to active Maleimide Groups
  • Specificity: Exposed thiol (SH) groups on biomolecule
  • Bead ligand: Chemically active Maleimide groups
  • Resulting bond: Irreversible Thioether
  • Protein binding capacity: ≥15 mg/ml
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Matrix: 6% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three months upon delivery, unopened
  • Chemical Structure: Maleimide structure


The resulting Thioether bond between the Maleimide MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Maleimide Activated MagBeads are also available as agarose resin if you prefer this method catalog or as XL MagBeads if you need higher flow rates for your downstream application.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR