PureCube HiCap StrepTactin® Compact Cartridge

Supplier: CUBE BIOTECH

34302 34304 34306 34308
77393-104EA 290.09 CAD
77393-104 77393-106 77393-108 77393-110
PureCube HiCap StrepTactin® Compact Cartridge
Protein Purification Protein Purification Reagents

HiCap StrepTactin® Compact Cartridge is a FPLC-ready column that is pre-packed with our PureCube HiCap StrepTactin® agarose resin.


  • Usage: Highly specific affinity purification of Strep-(WSHPQFEK)-tagged proteins
  • Specificity:Affinity to Strep-tagged (WSHPQFEK) proteins
  • Binding capacity: >5 mg/ml
  • Bead Ligand: StrepTactin®
  • Bead size: 100 μm
  • Matrix: 6% cross-linked agarose
  • Recommended flow rates: 0.25 to 1 ml/min
  • Max. flow rates: 6 ml/min
  • Filling quantity: Delivered as a 50% suspension
  • Chemical stability: 2% Tween 20, 2% Triton X-100, 2% IGEPAL® 630/Nonidet P40, n-Octyl-beta-D-glucopyranoside, 0,2% n-Nonyl-beta-D-glucopyranoside, 0,35% N-Decyl-beta-D-maltoside, 2% Lauryl-sarcosine, 0, 1% SDS, 0,3% CHAPS, 1M Guanidine HCl, 1mM PMSF, 10% Ethanol, 5M NaCl, 2M (NH4)SO₄, 1M CaCl₂, 25% Glycerol
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C


PureCube StrepTactin® agarose resin beads consist of 6% cross-linked agarose coupled to the StrepTactin protein.


Required equipment and materials:
-FPLC hardware like e.g. ÄKTA
-Binding Buffer
-Elution Buffer

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR