PureCube HiCap StrepTactin® MagBeads

Supplier: CUBE BIOTECH

34201 34205 34225 34290
77393-096EA 235.45 CAD
77393-096 77393-098 77393-100 77393-102
PureCube HiCap StrepTactin® MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube StrepTactin® MagBeads are a suspension of 6% cross-linked magnetic agarose beads coupled to a StrepTactin protein.


  • Usage: Highly specific affinity purification of Strep-(WSHPQFEK)-tagged proteins
  • Specificity: Affinity to Strep-tagged (WSHPQFEK) proteins
  • Binding capacity: >7 mg/ml
  • Bead ligand: StrepTactin®
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 5 % suspension
  • Chemical stability: 2% Tween 20, 2% Triton X-100, 2% IGEPAL® 630/Nonidet P40, n-Octyl-beta-D-glucopyranoside, 0,2% n-Nonyl-beta-D-glucopyranoside, 0,35% N-Decyl-beta-D-maltoside, 2% Lauryl-sarcosine, 0, 1% SDS, 0,3% CHAPS, 1M Guanidine HCl, 1mM PMSF, 10% Ethanol, 5M NaCl, 2M (NH4)SO₄, 1M CaCl₂, 25% Glycerol
  • Short-term storage: In equilibration buffer
  • Long-term storage: In 20% ethanol at 4 °C


PureCube StrepTactin® MagBeads enable protein yields of up to 7 mg protein per ml MagBeads and were developed for protein purification assays using the Strep-tag® and magnetic separation. PureCube StrepTactin® products are also obtainable as agarose resin. Furthermore, PureCube StrepTactin® MagBeads XL are also available for very vicious cell media.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR