PureCube IDA Agarose

Supplier: CUBE BIOTECH

30715 30701
77392-932PK 11522.97 CAD
77392-932 77392-922 77392-924 77392-926 77392-928 77392-930
PureCube IDA Agarose
Protein Purification Protein Purification Reagents

PureCube IDA (iminodiacetic acid) agarose resin was developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead ligand: IDA
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 40 µm and, thus, excellent mechanical stability. Since the affinity of an IDA-metal-ion-complex depends on the loaded metal ion, we offer IDA agarose resin uncoupled to any metal ion in case we do not offer your desired IDA + metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads. PureCube IDA resin is also available with a bead diameter of 400 µm for higher FPLC flow rates or as IDA magnetic beads. Furthermore, PureCube IDA agarose is also obtainable pre-packed into FPLC cartridges/columns.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR