PureCube Compact Cartridge Fe-NTA

Supplier: CUBE BIOTECH

31606-FE 31608-FE 31602-FE 31604-FE
77393-634EA 301.69 CAD
77393-634 77393-642 77393-618 77393-626
PureCube Compact Cartridge Fe-NTA
Protein Purification Protein Purification Reagents

PureCube Fe-NTA compact cartridge is an FPLC-ready column pre-packed with our PureCube Fe-NTA agarose resin.


  • Purity: >90% (Determined by SDS-PAGE)
  • Number of amino acids: 184
  • molecular mass: 21468 Da
  • Extinction coefficient (in water) ε280:19940 M⁻¹cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Origin: Human
  • Sequence (1-Letter Code): MGSSHHHHHH ENLYFQ GSTFSKLREQ LGPVTQEFWD NLEKETEGLR QEMSKDLEEV AKVQPYLDDF QKKWQEEMEL RQKVEPLGEE MRDRARAHVD ALRTHLAPYS DELRQRLAAR LEALKENGGA RLAEYHAKAT EHLSTLSEKA KPALEDLRQG LLPVLESFKV SFLSALEEYT KKLNTQ
  • Affinity tag: N-Terminal His-tag


PureCube Fe-NTA is used for the purification of His-tagged proteins and the enrichment of phosphorylated proteins. The PureCube cartridge/column that is used here is compatible with almost all commonly used FPLC systems like ÄKTA TM. As an alternative, we also offer PureCube Fe-NTA MagBeads, the more common format of Fe-NTA beads for phosphopeptide enrichment.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR