PureCube Compact Cartridge Cu-NTA

Supplier: Cube Biotech
PureCube
31606-CU 31602-CU 31604-CU 31608-CU
77393-616EA 100.9 CAD
77393-616 77393-624 77393-640 77393-632
PureCube Compact Cartridge Cu-NTA
Protein Purification Protein Purification Reagents
The PureCube Cu-NTA compact cartridge is an IMAC chromatography column prepacked with PureCube Cu-NTA resin. You can choose between two sizes, 1 ml bed volume and 5 ml bed volume.

  • Purity: >90% (determined by SDS-PAGE)
  • Number of amino acids: 217
  • Molecular mass: 25309 Da
  • Extinction coefficient (in water) ε280: 25440 M⁻¹ cm⁻¹
  • Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
  • Sequence (Human origin): MGSSHHHHHH ENLYFQ GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
  • Affinity tag: N-Terminal His-tag

Please have a look at the given features to ensure that the cartridge/column is compatible with your FPLC hardware like ÄKTA TM. If this is not the case we recommend buying the sole loose PureCube Cu-NTA resin and packing the columns yourself.

Alternatively, we also offer our PureCube Cu-NTA products as magnetic beads, if you prefer this IMAC method.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR