PureCube 100 NTA Agarose
Supplier: CUBE BIOTECH
PureCube NTA (nitrilotriacetic acid) agarose resin was developed for IMAC-based protein purification and protein enrichment methods. The beads in this suspension have an average diameter of 100 µm.
- Usage: Specific binding and purification of 6× His tagged proteins
- Specificity: Affinity to His tagged proteins
- Binding capacity: >100 mg/ml
- Bead ligand: Ni-INDIGO
- Bead size: 100 μm
- Matrix: 6% highly cross-linked agarose
- pH Stability: 2 to 13
- Chelator stability: Stable in buffer containing 20 mM DTT and 20 mM EDTA
Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA agarose resin uncoupled to any metal ion in case we do not offer your desired NTA + metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...