PureCube 100 Compact Cartridge Co-NTA
Supplier: CUBE BIOTECH
74602
74604
74606
74608
77393-464EA
129.56
CAD
77393-464
77393-466
77393-468
77393-470
PureCube 100 Compact Cartridge Co-NTA
Protein Purification
Protein Purification Reagents
The PureCube 100 Co-NTA compact cartridge is an IMAC chromatography column prepacked with PureCube 100 Co-NTA resin.
- Usage: Specific binding and purification of 6× His-tagged proteins
- Specificity: Affinity to His-tagged proteins
- Binding capacity: >100 mg/ml
- Bead ligand: Ni-INDIGO
- Bead size: 30 μm
- Filling quantity: Delivered as a 25% suspension
- Chelator stability: Stable in buffer containing 20 mM DTT and 20 mM EDTA
- pH stability: 2 to 14
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
You can choose between two sizes, 1 ml bed volume and 5 ml bed volume. The PureCube cartridge/column that is used here is compatible with almost all commonly used FPLC systems like ÄKTA.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...