PureCube 100 Co-NTA Agarose

Supplier: Cube Biotech

74403 74405 74410 74412 74415
77393-452EA 40.69 CAD
77393-452 77393-454 77393-456 77393-458 77393-460 77393-462
PureCube 100 Co-NTA Agarose
Protein Purification Protein Purification Reagents

Our PureCube 100 Co-NTA agarose resins are agarose resin beads with a diameter of 100 µm. They are used for the purification of active His-tagged proteins from cells.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead ligand: NTA
  • Bead size: 100 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


Our year-long experience in manufacturing agarose resin lead to the high yield of 30 mg protein per ml resin, which is leading in the market compared to other Co-NTA suppliers. PureCube Co-NTA resins are suited for batch spin columns and FPLC. Their diameter provides a great balance between mechanical stability and flow rates.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR