PureCube 100 Compact Cartridge Ni-NTA

Supplier: CUBE BIOTECH

74302 74304 74306 74308
77393-444EA 127.63 CAD
77393-444 77393-446 77393-448 77393-450
PureCube 100 Compact Cartridge Ni-NTA
Protein Purification Protein Purification Reagents

The PureCube 100 Ni-NTA compact cartridge is an IMAC chromatography column prepacked with PureCube 100 Ni-NTA resin.


  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead ligand: Co-NTA
  • Bead size: 100 μm
  • Filling quantity: Delivered as a 50% suspension


Required equipment and materials:
- FPLC hardware like e.g. ÄKTA
- Binding buffer
- Elution buffer


You can choose between two sizes, 1 ml bed volume and 5 ml bed volume. Please look at the given features table to ensure that the cartridge/column is compatible with your FPLC hardware like ÄKTA. If a compatibility cannot be ensured we recommend buying the sole loose PureCube 100 Ni-NTA resin and packing the columns yourself.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR