PureCube Epoxy Activated Agarose XL

Supplier: CUBE BIOTECH

58705 58710 58712 58703
77393-384EA 1498.03 CAD
77393-384 77393-386 77393-388 77393-382
PureCube Epoxy Activated Agarose XL
Protein Purification Protein Purification Reagents

PureCube epoxy agarose XL resin is your best option to couple biomolecules that carry thiol (SH) groups.


  • Usage: Coupling proteins to active maleimide groups
  • Specificity: Exposed thiol (SH) groups on biomolecule
  • Bead ligand: Chemically active Maleimide groups
  • Resulting bond: irreversible thioether
  • Protein binding capacity: ≥15 mg/ml
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • Matrix: 6% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 100 mM sodium hydrogen carbonate, 0.02% sodium azide, pH 7.5 at 4 °C or 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Shelf life: If properly stored - three months upon delivery, unopened
  • Chemical Structure: Maleimide structure


The resulting Thioether bond between the Epoxy agarose beads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube Epoxy activated agarose is also available as magnetic beads if you prefer this method or as agarose resin with a smaller bead diameter if your downstream application requires more mechanically stable beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR