PureCube Phenyl Agarose
Supplier: CUBE BIOTECH
39103
39105
39110
39112
39115
77393-124EA
162.14
CAD
77393-124
77393-126
77393-128
77393-130
77393-132
PureCube Phenyl Agarose
Protein Purification
Protein Purification Reagents
Our PureCube phenyl agarose resin is a suspension of purification beads for hydrophobic interaction chromatography.
- Antibody name: Rho-1d4-1
- Cell line/origin: Mouse, hybridoma
- Antibody purification: rProtein A
- Quantity: 200 µg
- Concentration: 4.11 mg/ml (by UV absorbance at 280 nm)
- Physical State: Liquid (sterile filtered)
- Color: Clear
- Buffer: 0.25 M sodium bicarbonate, 0.5 M sodium chloride, pH = 8.3
- Purity: >95% (determined by non-denaturing SDS-PAGE)
- Shipment temperature: Shipped on ice
- Short-term storage: Store vial at 4 °C. Centrifuge product if not completely clear after standing at room temperature and/or heating to 37 °C
- Long-term Storage: Aliquot contents and freeze at –20 °C or below. Avoid repeated freeze-thaw cycles
Hydrophobic Interaction Chromatography (HIC) is a reliable method to separate proteins and other biomolecules based on their hydrophobicity. If you use HIC with your sample for the first time we recommend testing all of our HIC resins before.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...