PureCube Butyl Agarose

Supplier: CUBE BIOTECH

39003 39005 39010 39012 39015
77393-112EA 162.14 CAD
77393-112 77393-114 77393-116 77393-118 77393-120
PureCube Butyl Agarose
Protein Purification Protein Purification Reagents

Our PureCube butyl agarose resin is a suspension of purification beads for hydrophobic interaction chromatography.


  • Usage: As a matrix for Hydrophobic Interaction Chromatography (HIC)
  • Specificity: Hydrophobic biomolecules and proteins
  • Bead size: 40 μm
  • Bead ligand: Phenyl groups
  • Phenyl group density: 40 µmol/ml
  • BSA binding capacity: 25 mg/ml
  • lysozyme binding capacity: 45 mg/ml
  • pH stability: 2 to 13
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% cross-linked agarose
  • Short term storage: In neutral buffer at 4 °C
  • Long term storage: 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
  • Chemical structure of phenyl ligand; phenyl group structure


Hydrophobic Interaction Chromatography (HIC) is a reliable method to separate proteins and other biomolecules based on their hydrophobicity. If you use HIC with your sample for the first time we recommend testing all of our HIC resins before.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR