PureCube StrepTactin® HiCap StrepTactin® Agarose
Supplier: CUBE BIOTECH
PureCube StrepTactin® agarose resin beads consist of 6% cross-linked agarose coupled to the streptactin protein.
- Usage: As a matrix for Hydrophobic Interaction Chromatography (HIC)
- Specificity: Hydrophobic biomolecules and proteins
- Bead size: 40 μm
- Bead ligand: Butyl groups
- Butyl group density: 40 µmol/ml
- BSA binding capacity: 25 mg/ml
- Lysozyme binding capacity: 45 mg/ml
- pH stability: 2 to 13
- Filling quantity: Delivered as a 50% suspension
- Matrix: 7.5% cross-linked agarose
- Short term storage: In neutral buffer at 4 °C
- Long term storage: 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
- Chemical structure of butyl ligand: Butyl group structure
PureCube StrepTactin® agarose resin beads enable protein yields of up to 5 mg protein per ml resin. They are intended for FPLC and batch spin-based protein purification. PureCube StrepTactin® agarose can also be purchased pre-packed in FPLC cartridges and columns. Furthermore, PureCube StrepTactin® agarose XL resin is also available for higher FPLC flow rates or very viscous cell media. Alternatively, Cube Biotech also offers PureCube StrepTactin® magnetic beads/MagBeads.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...