HighSpec HighSpec Rho1D4 MagBeads

Supplier: CUBE BIOTECH

33201 33205 33225
77393-072EA 727.56 CAD
77393-072 77393-074 77393-076
HighSpec HighSpec Rho1D4 MagBeads
Protein Purification Protein Purification Magnetic Beads

HighSpec Rho1D4 MagBeads are a suspension of 30 µm magnetic agarose beads coupled to an anti-Rho1D4 antibody.


  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead Ligand: Anti-Rho1D4 antibody
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Recommended flow rates: 0.25 to 1 ml/min
  • Max. flow rates: 6 ml/min
  • Filling quantity: Delivered as a 50% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C


Required equipment and materials:
- FPLC hardware like e.g. AKTA
- Binding buffer
- Elution buffer


HighSpec Rho1D4 MagBeads are meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. Additionally, we offer Rho1D4-tag affinity resin with a bead diameter of 90 µm for very viscous cell media. Alternatively, Rho1D4 Agarose resin is also available if you prefer this affinity chromatography method.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR