PureCube Ni-IDA MagBeads

Supplier: CUBE BIOTECH

30201 30205 30225 30290 30291
77392-904EA 129.31 CAD
77392-904 77392-906 77392-908 77392-910 77392-912
PureCube Ni-IDA MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube Ni-IDA MagBeads were developed by Cube Biotech for His-tag protein purification.


  • Usage: Specific binding and purification of Rho1D4-tagged proteins
  • Specificity: affinity to Rho1D4-tagged (TETSQVAPA) proteins
  • Binding capacity: >3 mg/ml
  • Bead ligand: anti-Rho1D4 antibody
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • Short-term storage: In neutral buffer at 4 °C
  • Long-term storage: In neutral buffer with 20% ethanol at 4 °C


Our year-long experience manufacturing agarose resin led to a high yield of 70 mg protein per ml resin, leading in the market compared to other Ni-IDA suppliers. Ni-IDA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin or as XL magnetic beads for more viscous cell media. For convenience, Cube Biotech also offers its own MagBead separator for use with our magnetic beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR