PureCube Zn-NTA MagBeads

Supplier: Cube Biotech

31501-ZN 31505-ZN 31525-ZN 31591-ZN 31590-ZN
77393-566EA 202.65 CAD
77393-566 77393-578 77393-590 77393-612 77393-602
PureCube Zn-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube Zn-NTA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.


  • Usage: Specific binding and purification of 6× His-tagged proteins
  • Binding of Zinc-binding proteins
  • Specificity: Affinity to His-tagged proteins
  • Affinity to Zinc-finger proteins
  • Binding capacity: >50 mg/ml
  • Bead Ligand: Zn-NTA
  • Bead size: 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Metal ion capacity: >15 µeqv Zn²⁺/ml
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


Our year-long experience in manufacturing agarose resin lead to the high yield of 50 mg protein per ml resin, which is leading in the market compared to other Ni-IDA suppliers. Ni-IDA products by Cube Biotech are also available as agarose resin beads for FPLC and batch spin. For your convenience Cube Biotech also offers its own MagBead separator for use with our magnetic beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR