PureCube Al-NTA Agarose
Supplier: CUBE BIOTECH
31403-AL
31405-AL
31410-AL
31412-AL
77393-518EA
39.06
CAD
77393-518
77393-526
77393-534
77393-542
77393-550
PureCube Al-NTA Agarose
Protein Purification
Protein Purification Reagents
PureCube Al-NTA agarose resin beads were developed for the enrichment of phosphorylated proteins.
- Usage: Enrichment of phosphopeptides
- Specificity: Affinity to phosphorylated biomolecules
- Bead ligand: Al-NTA
- Bead size: 40 µm
- Filling quantity: Delivered as a 50% suspension
- Matrix: 7.5% crosslinked agarose
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- pH stability: 2 to 14
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
PureCube Al-NTA agarose resin beads can also be used for the purification of His-tagged proteins as a secondary function. The affinity matrix is based on 7.5% cross-linked agarose that was magnetized. The bead ligand is Al-NTA and the particle diameter is 40 µm. They are very homogeneous in size, yielding a high degree of reproducibility.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...