PureCube Glutathione MagBeads XL

Supplier: CUBE BIOTECH

57105 57125 57190
77393-320EA 704.4 CAD
77393-320 77393-322 77393-324
PureCube Glutathione MagBeads XL
Protein Purification Protein Purification Magnetic Beads

The protein purification with the PureCube Glutathione MagBeads and the GST-tag exploit the specific enzyme-substrate binding to gain high levels of purity while maintaining a high protein yield.


  • Usage: Specific binding and purification of GST-tagged proteins
  • Specificity: Affinity to GST(Glutathione-S-Transferase)-tagged proteins
  • Binding capacity: >20 mg/ml
  • Bead Ligand: L-Glutathione (reduced)
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH Stability: 3 to 12
  • Stabilities: 1 M sodium acetate, pH 4.0, or 6 M guanidinehydrochloride, organic solvents (e.g., 70% (v/v) ethanol), 1% (w/v) SDS, 0.1 M NaOH, 0.1 M HCl


The pairing of an enzyme to its substrate is one of the most specific combinations in life sciences. This is why GST-tag purifications exploit this specificity to gain high purity levels while maintaining a high protein yield. PureCube Glutathione MagBeads XL is a suspension of 90 µm magnetic purification beads that are used for this purpose. Their small size provides high mechanical stability.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR