PureCube NTA MagBeads XL

Supplier: CUBE BIOTECH

55905 55925 55990
77393-298EA 953.34 CAD
77393-298 77393-300 77393-302
PureCube NTA MagBeads XL
Protein Purification Protein Purification Magnetic Beads

PureCube NTA (nitrilotriacetic acid) XL MagBeads were developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: NTA
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 90 µm. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA MagBeads uncoupled to any metal ion for the case that we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR