PureCube IDA MagBeads XL

Supplier: CUBE BIOTECH

55705 55725 55790
77393-284EA 527.28 CAD
77393-284 77393-286 77393-288
PureCube IDA MagBeads XL
Protein Purification Protein Purification Magnetic Beads

PureCube IDA (nitrilotriacetic acid) XL MagBeads were developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: IDA
  • Bead size: 90 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 90 µm. Since the affinity of an IDA-metal-ion-complex depends on the loaded metal ion, we offer IDA MagBeads uncoupled to any metal ion for the case that we do not offer your desired IDA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR