PureCube Co-NTA MagBeads XL

Supplier: CUBE BIOTECH

55505 55525 55590
77393-270EA 1024.76 CAD
77393-270 77393-272 77393-274
PureCube Co-NTA MagBeads XL
Protein Purification Protein Purification Magnetic Beads

PureCube Co-NTA XL magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.


  • Usage: Specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead Ligand: Co-NTA
  • Bead size: 90 μm
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


Our year-long experience in manufacturing agarose resin lead to the high yield of 30 mg protein per ml resin, which is leading in the market compared to other Co-NTA suppliers. PureCube Co-NTA resins are suited for batch spin columns and FPLC. Their huge diameter provides great flow rates for even the most vicious cell media.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR