PureCube Ni-IDA MagBeads XL
Supplier: CUBE BIOTECH
55205
55225
55290
77393-256EA
656.17
CAD
77393-256
77393-258
77393-260
PureCube Ni-IDA MagBeads XL
Protein Purification
Protein Purification Magnetic Beads
PureCube Ni-IDA magnetic beads/MagBeads were developed by Cube Biotech for His-tag protein purification.
- Usage: Specific binding and purification of 6× His-tagged proteins
- Specificity: Affinity to His-tagged proteins
- Binding capacity: >40 mg/ml
- Bead Ligand: Ni-IDA
- Bead size: 90 μm
- Filling quantity: Delivered as a 25% suspension
- pH stability: 2 to 14
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
Our year-long experience in manufacturing agarose resin lead to the high yield of 40 mg protein per ml MagBeads. The XL beads are meant for viscous media and cell debris.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...