HighSpec Compact Cartridge Rho1D4
Supplier: CUBE BIOTECH
33306
33308
33302
33304
77393-084EA
2101.4
CAD
77393-084
77393-086
77393-080
77393-082
HighSpec Compact Cartridge Rho1D4
Protein Purification
Protein Purification Reagents
The HighSpec Rho1D4 Compact cartridge is a pre-packed FPLC column containing our HighSpec Rho1D4 Agarose resin.
- Usage: Specific binding and purification of Rho1D4-tagged proteins
- Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
- Binding capacity: >3 mg/ml
- Bead Ligand: anti-Rho1D4 antibody
- Bead size: 40 μm
- Matrix: 7.5% cross-linked agarose
- Recommended flow rates: 0.25 to 1 ml/min
- Max. flow rates: 6 ml/min
- Filling quantity: Delivered as a 50% suspension
- Short-term storage: In neutral buffer at 4 °C
- Long-term storage: In neutral buffer with 20% ethanol at 4 °C
- Required equipment and materials:
- FPLC hardware like e.g. ÄKTA
- Binding Buffer
- Elution Buffer
The HighSpec Rho1D4 Compact Cartridge is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. You can also purchase the pre-packed Rho1D4 agarose as loose resin. Alternatively, Rho1D4 MagBeads are also available if you prefer this affinity chromatography method.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...