PureCube Compact Cartridge Glutathione

Supplier: Cube Biotech

32302 32304 32306 32308
77393-054EA 108.29 CAD
77393-054 77393-056 77393-058 77393-060
PureCube Compact Cartridge Glutathione
Protein Purification Protein Purification Reagents

The protein purification with the PureCube Glutathione agarose and the GST-tag exploit the specific enzyme-substrate binding to gain high levels of purity while maintaining a high protein yield.


  • Usage: Specific binding and purification of GST-tagged proteins
  • Specificity: Affinity to GST(Glutathione-S-Transferase)-tagged proteins
  • Binding capacity: >10 mg/ml
  • Bead Ligand: L-Glutathione (reduced)
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • pH Stability: 3 to 12
  • Required equipment and materials:
  • FPLC hardware like e.g. ÄKTA
  • Binding Buffer
  • Elution Buffer


The pairing of an enzyme to its substrate is one of the most specific combinations in life sciences. The PureCube cartridge/column used here is compatible with almost all commonly used FPLC systems like ÄKTA. The PureCube Glutathione Compact cartridge is an FPLC column pre-packed with PureCube Glutathione Agarose. Alternatively, Glutathione MagBeads are also available if you prefer this affinity chromatography approach. Additional support products for GST-tag purification assays are: L-glutathione (reduced), essential compound of the elution buffer, GST-tag antibodies for western blot detection.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR