PureCube Compact Cartridge NTA

Supplier: CUBE BIOTECH

31902 31904 31906 31908
77393-026EA 93.22 CAD
77393-026 77393-028 77393-030 77393-032
PureCube Compact Cartridge NTA
Protein Purification Protein Purification Reagents

The PureCube NTA (nitrilotriacetic acid) Compact Cartridge was developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: NTA
  • Bead size: 40 μm
  • Matrix: 7.5% cross-linked agarose
  • Filling quantity: Delivered as a 50% suspension
  • Recommended FPLC flow rates: 0.5 to 2 ml/min
  • Maximal FPLC flow rate: 6 ml/min
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in the pre-packed resin have an average diameter of 40 µm. This small size provides excellent mechanical stability. The PureCube NTA agarose resin is packed into our PureCube Compact Cartridge. The Compact Cartridge is compatible with the most commonly used FPLC systems like ÄKTA. Since the affinity of an NTA-metal-ion-complex depends on the loaded metal ion, we offer NTA agarose resin uncoupled to any metal ion in case we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach your desired metal ions to the agarose beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR