PureCube Co-NTA Agarose

Supplier: CUBE BIOTECH

31403 31405 31410 31412 31415
77392-978EA 39.06 CAD
77392-978 77392-980 77392-982 77392-984 77392-986 77392-988
PureCube Co-NTA Agarose
Protein Purification Protein Purification Reagents

Our PureCube Co-NTA agarose resins are tiny agarose resin beads with a diameter of 40 µm. They are used for the purification of active His-tagged proteins from cells.


  • Usage: Specific binding and purification of 6× His tagged proteins
  • Specificity: Affinity to His tagged proteins
  • Binding capacity: >30 mg/ml
  • Bead Ligand: Co-NTA
  • Bead size: 40 μm
  • Filling quantity: Delivered as a 50% suspension
  • Required equipment:
  • Lysis Buffer
  • Wash Buffer
  • Elution Buffer
  • Ice bath
  • Refrigerated centrifuge for 50 ml tube (min 10000×g)
  • 50 ml centrifuge tube
  • Micropipettor and Micropipetting tips
  • Disposable gravity flow columns with capped bottom outlet, 2 ml
  • pH meter
  • End-over-end shaker
  • SDS-PAGE buffers, reagents and equipment Optional: Western -Blot reagents and equipment


Our year-long experience in manufacturing agarose resin led to a high yield of 40 mg protein per ml resin, leading in the market compared to other Co-NTA suppliers. PureCube Co-NTA resins are suited for batch spin columns and FPLC. Their small diameter provides them with excellent mechanical stability. We recommend our Co-NTA agarose beads with 100 µm diameter for higher flow rates and, for extreme cases, XL-sized Co-NTA beads. Furthermore, these beads are also available pre-packed in a column/cartridge or as magnetic beads.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR