PureCube Ni-NTA MagBeads
Supplier: CUBE BIOTECH
31201
31205
31225
31290
77392-962EA
169.25
CAD
77392-962
77392-964
77392-966
77392-968
PureCube Ni-NTA MagBeads
Protein Purification
Protein Purification Magnetic Beads
PureCube Ni-NTA MagBeads were developed by Cube Biotech for His-tag protein purification.
- Usage: Specific binding and purification of 6× His-tagged proteins
- Specificity: Affinity to His-tagged proteins
- Binding capacity: >80 mg/ml
- Bead Ligand: Ni-NTA
- Bead size: 30 μm
- Filling quantity: Delivered as a 25% suspension
- pH stability: 2 to 14
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
Our year-long experience manufacturing agarose resin led to the high yield of 80 mg protein per ml resin, leading in the market compared to other Ni-NTA suppliers. Ni-NTA products by Cube Biotech are also available as agarose resin beads or as XL magnetic beads. For convenience, Cube Biotech also offers a MagBead separator for use with our magnetic beads.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...