PureCube IDA MagBeads

Supplier: CUBE BIOTECH

30801 30805 30825 30890
77392-934EA 117.17 CAD
77392-934 77392-936 77392-938 77392-940
PureCube IDA MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube IDA (iminodiacetic acid) MagBeads were developed for IMAC-based protein purification and protein enrichment methods.


  • Usage: For coupling of metal ions for corresponding purification and/or enrichment purposes
  • Specificity: Varies
  • Binding capacity: Depends on the loaded ion
  • Purity: Depends on the loaded ion
  • Bead Ligand: IDA
  • Bead size: 30 μm
  • Matrix: 6% cross-linked agarose
  • Filling quantity: Delivered as a 25% suspension
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The beads in this suspension have an average diameter of 30 µm and, thus, excellent mechanical stability. Since the affinity of an IDA-metal-ion-complex depends on the loaded metal ion, we offer IDA MagBeads uncoupled to any metal ion for the case that we do not offer your desired NTA and metal ion combination. Through our coupling protocol, you can, in turn, attach desired metal ions to the agarose beads. PureCube IDA MagBeads are also available with a bead diameter of 90 µm. PureCube IDA products are also obtainable as agarose resin for FPLC assays or batch spin approaches.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR