Cell-Free E. coli Lysate
Supplier: CUBE BIOTECH
21001
21003
77392-642EA
301.15
CAD
77392-642
77392-644
Cell-Free E. coli Lysate
Protein Purification
Protein Purification Reagents
Our E. coli cell-free lysates are the isolated protein purification machinery of bacterial cells. They create a high-yielding and adjustable cell-free protein expression system.
- Average yields: 1.3 to 5 mg/ml
- Volume per vial: 350 µl
The advantages of cell-free protein expression are numerous, among other things it enables a scientist to:
- For direct stabilization and co-translational integration of membrane proteins into nanodiscs
- Expression of otherwise toxic proteins