PureCube Compact Cartridges
Supplier: CUBE BIOTECH
16912 16914 16918 16919
77392-444EA 63.74 CAD
77392-444 77392-446 77392-448 77392-450
PureCube Compact Cartridges
Chromatography Columns
Empty PureCube Compact cartridges can be filled with your affinity resin of choice.
  • Max. Flow Rate: 6 ml/min
  • Body Material: Polypropylene
Empty PureCube Compact cartridges provide the same technical features as PureCube prepacked cartridges and compact cartridges. They can be filled with your affinity resin of choice. PureCube Cartridges are available in two sizes, 1 and 5 ml bed volume, and two column geometries. Made from robust materials, the body and end plugs are resistant to the most commonly used reagents. The void volume in each end plug is minimal because the fluid is introduced through a narrow flow path. Recommended operational pressure is up to 3 bar (42 psi).
Delivery information: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: −20 °C for several years. Storage of dissolved copolymer: 2 to 8 °C for several day.