N-Octyl-β-D-glucopyranoside ≥99.5%, Crystallization grade

Supplier: CUBE BIOTECH

Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside

16002 16003 16004
77392-312EA 403.47 CAD
77392-312 77392-314 77392-316
N-Octyl-β-D-glucopyranoside ≥99.5%, Crystallization grade
N-Octyl-β-D-glucopyranoside

Octylglucopyranoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer glucosides with chain lengths from C-8 to C-10, as well as octylthioglucoside.


Molecular Weight: 292.38 g/mol
Form: White crystalline powder
CMC (in H2O): 18-20 mM (0.53%)
Mild, non-ionic detergent

Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Melting Pt: 110
MDL Number: MFCD00063288
CAS Number: 29836-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=481bbb6afdc223b421edd05dc064b45a89f9d1e6&vid=712217a8d69ca9f56e9c6db07beac4230a0500b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR