Anti-BLZF1 Rabbit Polyclonal Antibody

Supplier: ProSci
22-216
76761-472EA 688.39 CAD
76761-472
Anti-BLZF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BLZF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR