Anti-SLC6A18 Rabbit Polyclonal Antibody

Supplier: ProSci
14-669
76751-920EA 688.39 CAD
76751-920
Anti-SLC6A18 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC6A18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR