Anti-S Recombinant Antibody

Supplier: ProSci
11-587
77134-534EA 943.86 CAD
77134-534
Anti-S Recombinant Antibody
Antibodies
Type: Primary
Antigen: S
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG1/kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR