Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: ProSci
16-689
76753-176EA 688.37 CAD
76753-176
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR